Anti-UBE2T Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302510EA 454 EUR
ORIGTA302510
Anti-UBE2T Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000 Western Blot: 01-03µg/ml

Type: Primary
Antigen: UBE2T (ubiquitin-conjugating enzyme E2T (putative))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR