Anti-MARK4 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302535EA 454 EUR
ORIGTA302535
Anti-MARK4 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:128000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: MARK4 (MAP/microtubule affinity-regulating kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR