Anti-MDM2 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302536EA 547 EUR
ORIGTA302536
Anti-MDM2 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-10µg/ml

Type: Primary
Antigen: MDM2 (MDM2 oncogene, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR