Anti-NCF4 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302546EA 547 EUR
ORIGTA302546
Anti-NCF4 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 03-13µg/ml

Type: Primary
Antigen: NCF4 (neutrophil cytosolic factor 4, 40kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR