Anti-NOVA1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302553EA 550 EUR
ORIGTA302553
Anti-NOVA1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 001-003µg/ml

Type: Primary
Antigen: NOVA1 (neuro-oncological ventral antigen 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR