Anti-OGT Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302556EA 547 EUR
ORIGTA302556
Anti-OGT Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.05-0.2µg/ml.An

Type: Primary
Antigen: OGT (O-linked N-acetylglucosamine (GlcNAc) transferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR