Anti-PCBP4 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302562EA 454 EUR
ORIGTA302562
Anti-PCBP4 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:128000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: PCBP4 (poly(rC) binding protein 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR