Anti-PCK2 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302563EA 547 EUR
ORIGTA302563
Anti-PCK2 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:8000; Western Blot: 05-10 µg/ml

Type: Primary
Antigen: PCK2 (phosphoenolpyruvate carboxykinase 2 (mitochondrial))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR