Anti-PEBP1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302567EA 454 EUR
ORIGTA302567
Anti-PEBP1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 001-003µg/ml

Type: Primary
Antigen: PEBP1 (phosphatidylethanolamine binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR