Anti-ANP32B Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302570EA 547 EUR
ORIGTA302570
Anti-ANP32B Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.1-0.3?g/ml; Immunochemistry: 3-5?g/ml; ELISA: 1:16000

Type: Primary
Antigen: ANP32B (acidic (leucine-rich) nuclear phosphoprotein 32 family, member B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR