Anti-SAR1B Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302583EA 454 EUR
ORIGTA302583
Anti-SAR1B Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:8000; Western Blot: 003-01µg/ml

Type: Primary
Antigen: SAR1B (secretion associated, Ras related GTPase 1B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR