Anti-PDHX Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302585EA 547 EUR
ORIGTA302585
Anti-PDHX Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 01-1µg/ml; Immunochemistry: 1-3µg/ml

Type: Primary
Antigen: PDHX (pyruvate dehydrogenase complex, component X)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR