Anti-SAE1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302596EA 547 EUR
ORIGTA302596
Anti-SAE1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:8000; Western Blot: 1µg/ml

Type: Primary
Antigen: SAE1 (SUMO1 activating enzyme subunit 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR