Anti-HTR1B Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302601EA 500 EUR
ORIGTA302601
Anti-HTR1B Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:8000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: HTR1B (5-hydroxytryptamine (serotonin) receptor 1B, G protein-coupled)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR