Anti-ERP44 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302627EA 500 EUR
ORIGTA302627
Anti-ERP44 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 02-06µg/ml

Type: Primary
Antigen: ERP44 (endoplasmic reticulum protein 44)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR