Anti-VDAC2 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302629EA 550 EUR
ORIGTA302629
Anti-VDAC2 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml; ELISA: 1:4000

Type: Primary
Antigen: VDAC2 (voltage-dependent anion channel 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR