Anti-WNT3 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302636EA 454 EUR
ORIGTA302636
Anti-WNT3 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 1-3 ?g/ml; Immunochemistry: 2-4 ?g/ml

Type: Primary
Antigen: WNT3 (wingless-type MMTV integration site family, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR