Anti-CAV1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302639EA 500 EUR
ORIGTA302639
Anti-CAV1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:8000; Western Blot: 0.01-0.03?g/ml

Type: Primary
Antigen: CAV1 (caveolin 1, caveolae protein, 22kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR