Anti-BAK1 Goat polyclonal antibody

Supplier: OriGene
ORIGTA302647EA 454 EUR
ORIGTA302647
Anti-BAK1 Goat polyclonal antibody
Antibodies
Recommended Dilutions: Western Blot: 05-1µg/ml

Type: Primary
Antigen: BAK1 (BCL2-antagonist/killer 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR