Anti-CDKN2A Goat polyclonal antibody

Supplier: OriGene
ORIGTA302655EA 547 EUR
ORIGTA302655
Anti-CDKN2A Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 01-1µg/ml

Type: Primary
Antigen: CDKN2A (cyclin-dependent kinase inhibitor 2A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR