Anti-NQO1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302678EA 454 EUR
ORIGTA302678
Anti-NQO1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:2000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: NQO1 (NAD(P)H dehydrogenase, quinone 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR