Anti-PARK7 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302679EA 547 EUR
ORIGTA302679
Anti-PARK7 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000 Western Blot: 05-1µg/ml

Type: Primary
Antigen: PARK7 (parkinson protein 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR