Anti-XBP1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302701EA 547 EUR
ORIGTA302701
Anti-XBP1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:4000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: XBP1 (X-box binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR