Anti-BCL7A Goat polyclonal antibody

Supplier: OriGene
ORIGTA302703EA 550 EUR
ORIGTA302703
Anti-BCL7A Goat polyclonal antibody
Antibodies
Recommended Dilutions: Western Blot: 05-2µg/ml

Type: Primary
Antigen: BCL7A (B-cell CLL/lymphoma 7A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR