Anti-PPP1R15A Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302708EA 550 EUR
ORIGTA302708
Anti-PPP1R15A Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:128000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: PPP1R15A (protein phosphatase 1, regulatory subunit 15A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR