Anti-MTR Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302710EA 547 EUR
ORIGTA302710
Anti-MTR Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml; Immunochemistry: 3-5?g/ml; ELISA: 1:32000

Type: Primary
Antigen: MTR (5-methyltetrahydrofolate-homocysteine methyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR