Anti-CDKN1B Goat polyclonal antibody

Supplier: OriGene
ORIGTA302712EA 454 EUR
ORIGTA302712
Anti-CDKN1B Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: CDKN1B (cyclin-dependent kinase inhibitor 1B (p27, Kip1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR