Anti-SLC16A7 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302722EA 550 EUR
ORIGTA302722
Anti-SLC16A7 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:128000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: SLC16A7 (solute carrier family 16 (monocarboxylate transporter), member 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR