Anti-BAG2 Goat polyclonal antibody

Supplier: OriGene
ORIGTA302728EA 547 EUR
ORIGTA302728
Anti-BAG2 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:8000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: BAG2 (BCL2-associated athanogene 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR