Anti-ARFGAP3 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302737EA 547 EUR
ORIGTA302737
Anti-ARFGAP3 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: ARFGAP3 (ADP-ribosylation factor GTPase activating protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR