Anti-PPP2R5D Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302740EA 547 EUR
ORIGTA302740
Anti-PPP2R5D Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: PPP2R5D (protein phosphatase 2, regulatory subunit B', delta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR