Anti-DUSP16 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302742EA 500 EUR
ORIGTA302742
Anti-DUSP16 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000 Western Blot: 2-4µg/ml

Type: Primary
Antigen: DUSP16 (dual specificity phosphatase 16)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR