Anti-NOS1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302749EA 454 EUR
ORIGTA302749
Anti-NOS1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.1-0.3 ?g/ml

Type: Primary
Antigen: NOS1 (nitric oxide synthase 1 (neuronal))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR