Anti-PACSIN3 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302752EA 502 EUR
ORIGTA302752
Anti-PACSIN3 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: PACSIN3 (protein kinase C and casein kinase substrate in neurons 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR