Anti-DDAH2 Goat polyclonal antibody

Supplier: OriGene
ORIGTA302753EA 454 EUR
ORIGTA302753
Anti-DDAH2 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:2000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: DDAH2 (dimethylarginine dimethylaminohydrolase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR