Anti-NUMB Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302758EA 547 EUR
ORIGTA302758
Anti-NUMB Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 03-10 µg/ml

Type: Primary
Antigen: NUMB (numb homolog (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR