Anti-FXYD5 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302772EA 454 EUR
ORIGTA302772
Anti-FXYD5 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 05-15µg/ml

Type: Primary
Antigen: FXYD5 (FXYD domain containing ion transport regulator 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR