Anti-TFEB Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302788EA 547 EUR
ORIGTA302788
Anti-TFEB Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 0.5-1.5 ?g/ml; Immunochemistry: 2-4 ?g/ml

Type: Primary
Antigen: TFEB (transcription factor EB)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR