Anti-TFEC Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302789EA 547 EUR
ORIGTA302789
Anti-TFEC Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 05-2µg/ml

Type: Primary
Antigen: TFEC (transcription factor EC)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR