Anti-MXD4 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302831EA 454 EUR
ORIGTA302831
Anti-MXD4 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 05-2µg/ml

Type: Primary
Antigen: MXD4 (MAX dimerization protein 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR