Anti-SART1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302845EA 547 EUR
ORIGTA302845
Anti-SART1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 003-01µg/ml

Type: Primary
Antigen: SART1 (squamous cell carcinoma antigen recognized by T cells)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR