Anti-CDYL Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302848EA 550 EUR
ORIGTA302848
Anti-CDYL Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:4000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: CDYL (chromodomain protein, Y-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR