Anti-ARIH1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302850EA 547 EUR
ORIGTA302850
Anti-ARIH1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 02-1µg/ml

Type: Primary
Antigen: ARIH1 (ariadne RBR E3 ubiquitin protein ligase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR