Anti-MID2 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302853EA 454 EUR
ORIGTA302853
Anti-MID2 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3µg/ml; Immunochemistry: 5-10µg/ml

Type: Primary
Antigen: MID2 (midline 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR