Anti-RNF141 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302869EA 547 EUR
ORIGTA302869
Anti-RNF141 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 1µg/ml

Type: Primary
Antigen: RNF141 (ring finger protein 141)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR