Anti-USP11 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302871EA 550 EUR
ORIGTA302871
Anti-USP11 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:8000 Western Blot: 1µg/ml

Type: Primary
Antigen: USP11 (ubiquitin specific peptidase 11)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR