Anti-ACAP1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302896EA 550 EUR
ORIGTA302896
Anti-ACAP1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 05-2µg/ml

Type: Primary
Antigen: ACAP1 (ArfGAP with coiled-coil, ankyrin repeat and PH domains 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR