Anti-VPS45 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302901EA 454 EUR
ORIGTA302901
Anti-VPS45 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: VPS45 (vacuolar protein sorting 45 homolog (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR