Anti-PLEK Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302907EA 456 EUR
ORIGTA302907
Anti-PLEK Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 005-1µg/ml

Type: Primary
Antigen: PLEK (pleckstrin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR