Anti-MPP5 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302911EA 550 EUR
ORIGTA302911
Anti-MPP5 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3µg/ml; Immunochemistry: 5-10µg/ml

Type: Primary
Antigen: MPP5 (membrane protein, palmitoylated 5 (MAGUK p55 subfamily member 5))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR